Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
Rab5b
Entrez Gene ID
288779
Gene Name
RAB5B, member RAS oncogene family
Gene Aliases
-
Chromosome Location
Chr.7: 3104530 - 3123377 on Build Rnor_6.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001079936.1 NP_001073405.1 3 - 4 585 76
XM_006240750.2 XP_006240812.1 3 - 4 508 76
XM_006240751.2 XP_006240813.1 4 - 5 606 76
XM_006240752.2 XP_006240814.1 3 - 4 733 76
GenBank mRNA BC129101.1 - 3 - 4 585 76
FQ211489.1 - 3 - 4 723 76

View Full Product Details