Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
Slc22a3
Entrez Gene ID
29504
Gene Name
solute carrier family 22 (organic cation transporter), member 3
Gene Aliases
-
Chromosome Location
Chr.1: 48433079 - 48521261 on Build Rnor_6.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_019230.1 NP_062103.1 3 - 4 1063 95
GenBank mRNA AF055286.1 - 3 - 4 1063 95

View Full Product Details