The assay probe spans an exon junction. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001108223.1 | NP_001101693.1 | 3 - 4 | 661 | 90 |
XM_006244329.1 | XP_006244391.1 | 3 - 4 | 2750 | 90 | |
GenBank mRNA | BC158646.1 | - | 3 - 4 | 603 | 90 |