Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
Uhrf1
Entrez Gene ID
316129
Gene Name
ubiquitin-like with PHD and ring finger domains 1
Gene Aliases
Ac2-121, Np95
Chromosome Location
Chr.9: 10739306 - 10758403 on Build Rnor_6.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001008882.1 NP_001008882.1 4 - 5 602 84
GenBank mRNA AY321334.1 - 4 - 5 602 84

View Full Product Details