Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
Npepo
Entrez Gene ID
290963
Gene Name
aminopeptidase O
Gene Aliases
Aopep, Apo, RGD1309592
Chromosome Location
Chr.17: 507359 - 825062 on Build Rnor_6.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_008771431.1 XP_008769653.1 1 - 2 155 90

View Full Product Details