Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
Slc18a1
Entrez Gene ID
25693
Gene Name
solute carrier family 18 (vesicular monoamine transporter), member 1
Gene Aliases
CGAT, VMAT-1
Chromosome Location
Chr.16: 22362002 - 22395183 on Build Rnor_6.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_013152.2 NP_037284.2 3 - 4 718 130
XM_006252993.2 XP_006253055.1 2 - 3 918 130
XM_008771067.1 XP_008769289.1 2 - 3 918 130
GenBank mRNA M97380.1 - 3 - 4 747 130

View Full Product Details