Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
Angptl4
Entrez Gene ID
362850
Gene Name
angiopoietin-like 4
Gene Aliases
-
Chromosome Location
Chr.7: 18627814 - 18634043 on Build Rnor_6.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_199115.2 NP_954546.1 1 - 2 456 69
GenBank mRNA AY393999.1 - 1 - 2 316 69
BC078944.1 - 1 - 2 456 69

View Full Product Details