Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
Ssr4
Entrez Gene ID
29435
Gene Name
signal sequence receptor, delta
Gene Aliases
-
Chromosome Location
Chr.X: 156995763 - 156999709 on Build Rnor_6.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_017199.1 NP_058895.1 4 - 5 512 99
XM_006229571.2 XP_006229633.1 4 - 5 568 99
GenBank mRNA BC058482.1 - 4 - 5 425 99
FQ221421.1 - 2 - 3 128 99
FQ221949.1 - 2 - 3 128 99
FQ228311.1 - 2 - 3 128 99
FQ228501.1 - 2 - 3 128 99
Z19087.1 - 4 - 5 512 99

View Full Product Details