Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
Ccpg1
Entrez Gene ID
363098
Gene Name
cell cycle progression 1
Gene Aliases
-
Chromosome Location
Chr.8: 79660634 - 79692044 on Build Rnor_6.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001108770.1 NP_001102240.1 2 - 3 164 71
XM_006243332.2 XP_006243394.1 2 - 3 148 71
XM_006243337.2 XP_006243399.1 2 - 3 187 71
XM_006243341.2 XP_006243403.1 2 - 3 187 71
XM_008766315.1 XP_008764537.1 2 - 3 408 71
XM_008766316.1 XP_008764538.1 2 - 3 261 71
XM_008766317.1 XP_008764539.1 2 - 3 148 71
XM_008766318.1 XP_008764540.1 2 - 3 148 71
XM_008766319.1 XP_008764541.1 2 - 3 187 71
XM_008766320.1 XP_008764542.1 2 - 3 148 71

View Full Product Details