Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
Nlrp12
Entrez Gene ID
292541
Gene Name
NLR family, pyrin domain containing 12
Gene Aliases
Nalp12
Chromosome Location
Chr.1: 64506783 - 64538745 on Build Rnor_6.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001169142.1 NP_001162613.1 7 - 8 2737 72
XM_008758753.1 XP_008756975.1 7 - 8 2904 72

View Full Product Details