Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
Cacng6
Entrez Gene ID
140727
Gene Name
calcium channel, voltage-dependent, gamma subunit 6
Gene Aliases
-
Chromosome Location
Chr.1: 64307684 - 64322124 on Build Rnor_6.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_008758749.1 XP_008756971.1 2 - 3 1485 59
GenBank mRNA AF361344.1 - 2 - 3 405 59

View Full Product Details