The assay probe spans an exon junction. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_133387.2 | NP_596878.2 | 8 - 9 | 1252 | 83 |
XM_006255855.1 | XP_006255917.1 | 7 - 8 | 1261 | 83 | |
GenBank mRNA | AF374406.1 | - | 6 - 7 | 1093 | 83 |
BC085923.1 | - | 8 - 9 | 1252 | 83 |