Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
Vsig2
Entrez Gene ID
300520
Gene Name
V-set and immunoglobulin domain containing 2
Gene Aliases
RGD1561274
Chromosome Location
Chr.8: 40009691 - 40014734 on Build Rnor_6.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001106812.1 NP_001100282.1 4 - 5 710 91

View Full Product Details