Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
Copz1
Entrez Gene ID
315345
Gene Name
coatomer protein complex, subunit zeta 1
Gene Aliases
-
Chromosome Location
Chr.7: 144900091 - 144926299 on Build Rnor_6.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001108117.1 NP_001101587.1 1 - 2 69 96

View Full Product Details