Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
Tarbp2
Entrez Gene ID
363006
Gene Name
TAR (HIV-1) RNA binding protein 2
Gene Aliases
-
Chromosome Location
Chr.7: 144121731 - 144126860 on Build Rnor_6.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001034941.1 NP_001030113.1 3 - 4 428 89
XM_006242428.2 XP_006242490.1 3 - 4 508 89
GenBank mRNA BC104690.1 - 3 - 4 428 89
FQ228813.1 - 3 - 4 422 89

View Full Product Details