Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
Wnt10b
Entrez Gene ID
315294
Gene Name
wingless-type MMTV integration site family, member 10B
Gene Aliases
-
Chromosome Location
Chr.7: 140448465 - 140454268 on Build Rnor_6.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001108111.1 NP_001101581.1 2 - 3 407 65

View Full Product Details