The assay probe spans an exon junction. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001191710.1 | NP_001178639.1 | 6 - 7 | 2268 | 83 |
XM_006233082.2 | XP_006233144.1 | 6 - 7 | 1967 | 83 |