Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
Esco2
Entrez Gene ID
691979
Gene Name
establishment of sister chromatid cohesion N-acetyltransferase 2
Gene Aliases
-
Chromosome Location
Chr.15: 42501379 - 42518925 on Build Rnor_6.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_003751495.3 XP_003751543.1 10 - 11 1853 61

View Full Product Details