Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
Macc1
Entrez Gene ID
314539
Gene Name
metastasis associated in colon cancer 1
Gene Aliases
7a5, RGD1309838
Chromosome Location
Chr.6: 147315254 - 147392530 on Build Rnor_6.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001191775.1 NP_001178704.1 3 - 4 2334 111
XM_006240694.2 XP_006240756.1 4 - 5 2673 111
XM_008765003.1 XP_008763225.1 5 - 6 3112 111

View Full Product Details