Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
Adamtsl5
Entrez Gene ID
314626
Gene Name
ADAMTS-like 5
Gene Aliases
Thsd6
Chromosome Location
Chr.7: 12228723 - 12237382 on Build Rnor_6.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001108071.1 NP_001101541.1 7 - 8 747 117
XM_006240915.2 XP_006240977.1 9 - 10 1676 117
XM_006240916.2 XP_006240978.1 9 - 10 1666 117
XM_006240917.2 XP_006240979.1 9 - 10 1673 117
XM_006240918.2 XP_006240980.1 9 - 10 1616 117
XM_006240919.2 XP_006240981.1 9 - 10 1675 117
XM_006240920.2 XP_006240982.1 8 - 9 1505 117
GenBank mRNA FQ224249.1 - 9 - 10 976 117

View Full Product Details