Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
Vom2r53
Entrez Gene ID
286915
Gene Name
vomeronasal 2 receptor, 53
Gene Aliases
RGD1310577
Chromosome Location
Chr.7: 13151592 - 13184258 on Build Rnor_6.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_173131.2 NP_775154.1 2 - 3 524 131
GenBank mRNA AF016178.1 - 2 - 3 524 131

View Full Product Details