The assay probe spans an exon junction. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_138509.3 | NP_612518.2 | 6 - 7 | 830 | 121 |
XM_006235294.2 | XP_006235356.1 | 6 - 7 | 813 | 121 | |
XM_006235295.2 | XP_006235357.1 | 6 - 7 | 883 | 121 | |
GenBank mRNA | BC081726.1 | - | 6 - 7 | 830 | 121 |