The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001101005.1 | NP_001094475.1 | 1 - 2 | 187 | 111 |
GenBank mRNA | BC083575.1 | - | 1 - 2 | 187 | 111 |