Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
Tgif2
Entrez Gene ID
499929
Gene Name
TGFB-induced factor homeobox 2
Gene Aliases
RGD1560115, RGD1564927, Tgif2l1
Chromosome Location
Chr.3: 152906960 - 152924380 on Build Rnor_6.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001134983.1 NP_001128455.1 1 - 2 183 72
XM_006235427.2 XP_006235489.1 1 - 2 269 72
GenBank mRNA BC168719.1 - 1 - 2 183 72
FQ235168.1 - 1 - 2 121 72

View Full Product Details