The assay probe spans an exon junction. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001100885.1 | NP_001094355.1 | 2 - 3 | 362 | 87 |
XM_006235401.1 | XP_006235463.1 | 3 - 4 | 478 | 87 | |
XM_006235402.2 | XP_006235464.1 | 3 - 4 | 492 | 87 | |
GenBank mRNA | BC158548.1 | - | 3 - 4 | 441 | 87 |
FQ215756.1 | - | 3 - 4 | 415 | 87 | |
FQ220245.1 | - | 3 - 4 | 419 | 87 | |
FQ220303.1 | - | 3 - 4 | 419 | 87 | |
FQ220687.1 | - | 3 - 4 | 421 | 87 | |
FQ227515.1 | - | 3 - 4 | 422 | 87 | |
FQ229420.1 | - | 3 - 4 | 419 | 87 | |
FQ229727.1 | - | 3 - 4 | 422 | 87 | |
FQ229762.1 | - | 3 - 4 | 421 | 87 | |
FQ229855.1 | - | 3 - 4 | 455 | 87 | |
FQ230056.1 | - | 3 - 4 | 422 | 87 | |
FQ230407.1 | - | 3 - 4 | 435 | 87 | |
FQ231493.1 | - | 3 - 4 | 435 | 87 | |
FQ231820.1 | - | 3 - 4 | 454 | 87 | |
FQ232091.1 | - | 3 - 4 | 415 | 87 | |
S77900.1 | - | 2 - 3 | 362 | 87 |