Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
Cdh8
Entrez Gene ID
84408
Gene Name
cadherin 8
Gene Aliases
-
Chromosome Location
Chr.19: 6031935 - 6423809 on Build Rnor_6.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_053393.2 NP_445845.2 11 - 12 2435 79
XM_008772244.1 XP_008770466.1 10 - 11 1782 79
GenBank mRNA AB010436.1 - 11 - 12 2435 79

View Full Product Details