Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
Cdh8
Entrez Gene ID
84408
Gene Name
cadherin 8
Gene Aliases
-
Chromosome Location
Chr.19: 6031935 - 6423809 on Build Rnor_6.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_053393.2 NP_445845.2 9 - 10 2072 76
XM_008772244.1 XP_008770466.1 8 - 9 1419 76
GenBank mRNA AB010436.1 - 9 - 10 2072 76

View Full Product Details