Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
Cdh11
Entrez Gene ID
84407
Gene Name
cadherin 11
Gene Aliases
-
Chromosome Location
Chr.19: 2393059 - 2551245 on Build Rnor_6.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_053392.1 NP_445844.1 12 - 13 2369 61
XM_006255059.2 XP_006255121.1 12 - 13 4449 61

View Full Product Details