Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
Hmox1
Entrez Gene ID
24451
Gene Name
heme oxygenase (decycling) 1
Gene Aliases
HEOXG, Heox, Hmox, Ho-1, Ho1, hsp32
Chromosome Location
Chr.19: 14508634 - 14515455 on Build Rnor_6.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_012580.2 NP_036712.1 1 - 2 139 62
GenBank mRNA BC091164.1 - 1 - 2 139 62

View Full Product Details