Assay Design
The assay probe spans an exon junction.
Gene Transcripts
- Gene Symbol
- Hey2
- Entrez Gene ID
-
155430
- Gene Name
- hes-related family bHLH transcription factor with YRPW motif 2
- Gene Aliases
-
Herp1
- Chromosome Location
-
Chr.1: 29191170 - 29201257 on Build Rnor_6.0
Interrogated Sequence |
Translated Protein |
Exon Boundary |
Assay Location |
Amplicon Length |
RefSeq |
NM_130417.1 |
NP_569101.1 |
4 - 5 |
385 |
148 |
View Full Product Details