Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
Irx6
Entrez Gene ID
307715
Gene Name
iroquois homeobox 6
Gene Aliases
RGD1564830
Chromosome Location
Chr.19: 15726654 - 15733412 on Build Rnor_6.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001102413.1 NP_001095883.1 1 - 2 780 60
XM_006255173.2 XP_006255235.1 1 - 2 759 60

View Full Product Details