Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
Gpt2
Entrez Gene ID
307759
Gene Name
glutamic pyruvate transaminase (alanine aminotransferase) 2
Gene Aliases
ALT2, Cc2-5
Chromosome Location
Chr.19: 22599003 - 22633562 on Build Rnor_6.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001012057.1 NP_001012057.1 4 - 5 601 142
XM_008772405.1 XP_008770627.1 5 - 6 722 142
XM_008772406.1 XP_008770628.1 5 - 6 911 142
XM_008772407.1 XP_008770629.1 5 - 6 648 142
GenBank mRNA FQ223845.1 - - 797 142

View Full Product Details