The assay probe spans an exon junction. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_032079.2 | NP_114468.2 | 2 - 3 | 153 | 111 |
GenBank mRNA | BC087010.1 | - | 2 - 3 | 153 | 111 |
U95727.1 | - | 2 - 3 | 187 | 111 |