Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
Fbxo2
Entrez Gene ID
85273
Gene Name
F-box protein 2
Gene Aliases
-
Chromosome Location
Chr.5: 164972162 - 164977912 on Build Rnor_6.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_053511.1 NP_445963.1 1 - 2 87 88
XM_008764298.1 XP_008762520.1 2 - 3 290 88
GenBank mRNA AF098301.1 - 1 - 2 87 88
FQ214021.1 - 1 - 2 80 88

View Full Product Details