The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_153306.1 | NP_695218.1 | 8 - 9 | 1290 | 96 |
GenBank mRNA | AB036990.1 | - | 8 - 9 | 1290 | 96 |