The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_198130.1 | NP_937763.1 | 1 - 2 | 416 | 88 |
XM_006244087.2 | XP_006244149.1 | 2 - 3 | 763 | 88 | |
GenBank mRNA | AY191270.1 | - | 1 - 2 | 416 | 88 |
BC097266.1 | - | 1 - 2 | 356 | 88 |