Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
Myd88
Entrez Gene ID
301059
Gene Name
myeloid differentiation primary response 88
Gene Aliases
-
Chromosome Location
Chr.8: 128022512 - 128027461 on Build Rnor_6.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_198130.1 NP_937763.1 5 - 6 996 66
XM_006244087.2 XP_006244149.1 6 - 7 1343 66
GenBank mRNA AY191270.1 - 5 - 6 996 66
BC097266.1 - 5 - 6 936 66

View Full Product Details