The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_023103.1 | NP_075591.1 | 17 - 18 | 2148 | 112 |
XM_006237282.2 | XP_006237344.1 | 17 - 18 | 2188 | 112 | |
GenBank mRNA | M22359.1 | - | - | 1018 | 112 |
X52984.1 | - | 17 - 18 | 2148 | 112 |
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | XM_008763278.1 | XP_008761500.1 | 15 - 16 | 1860 | 112 |