Assay Design

The assay is designed to a sequence that does not exhibit typical exon-intron-exon gene structure, such as a fusion transcript, or an artificial, prokaryotic or mitochondrial sequence.

Gene Transcripts

Gene Symbol
Tmsb4x
Entrez Gene ID
81814
Gene Name
thymosin beta 4, X-linked
Gene Aliases
-
Chromosome Location
Chr.X: 28593405 - 28617267 on Build Rnor_6.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_031136.1 NP_112398.1 - 589 142
GenBank mRNA M34043.1 - - 589 142

View Full Product Details