Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
Pax8
Entrez Gene ID
81819
Gene Name
paired box 8
Gene Aliases
-
Chromosome Location
Chr.3: 1527316 - 1586023 on Build Rnor_6.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_031141.2 NP_112403.2 10 - 11 1371 64
XM_006233646.2 XP_006233708.1 10 - 11 2471 64
XM_008761610.1 XP_008759832.1 9 - 10 2807 64
XM_008761611.1 XP_008759833.1 8 - 9 2601 64
XM_008761612.1 XP_008759834.1 7 - 8 2535 64
XM_008761613.1 XP_008759835.1 8 - 9 2596 64
GenBank mRNA BC081779.1 - 10 - 11 1371 64
FQ221890.1 - 2 - 3 220 64
X94246.1 - 9 - 10 1254 64

View Full Product Details