Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
Coil
Entrez Gene ID
50998
Gene Name
coilin
Gene Aliases
-
Chromosome Location
Chr.10: 76320676 - 76336169 on Build Rnor_6.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_017360.1 NP_059056.1 1 - 2 246 60
GenBank mRNA AF220424.1 - 1 - 2 246 60
AF399643.1 - 1 - 2 246 60

View Full Product Details