Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
Fdxr
Entrez Gene ID
79122
Gene Name
ferredoxin reductase
Gene Aliases
AR
Chromosome Location
Chr.10: 103817724 - 103826413 on Build Rnor_6.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_024153.1 NP_077067.1 7 - 8 750 60
GenBank mRNA BC088844.1 - 7 - 8 757 60
D63761.1 - 7 - 8 750 60

View Full Product Details