The assay probe spans an exon junction. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_053291.3 | NP_445743.2 | 9 - 10 | 1120 | 154 |
GenBank mRNA | BC063161.1 | - | 9 - 10 | 1201 | 154 |
BC087651.1 | - | 9 - 10 | 1120 | 154 | |
FQ224830.1 | - | 4 - 5 | 613 | 154 |