The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_080697.2 | NP_542428.1 | 2 - 3 | 385 | 91 |
XM_006247077.2 | XP_006247139.1 | 2 - 3 | 164 | 91 | |
XM_008767940.1 | XP_008766162.1 | 2 - 3 | 155 | 91 | |
GenBank mRNA | AY034383.1 | - | 2 - 3 | 279 | 91 |
BC061874.1 | - | 2 - 3 | 385 | 91 |