Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
Tsks
Entrez Gene ID
292890
Gene Name
testis-specific serine kinase substrate
Gene Aliases
Stk22s1
Chromosome Location
Chr.1: 100926701 - 100953950 on Build Rnor_6.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_199088.1 NP_954519.1 8 - 9 1360 71
XM_006229014.2 XP_006229076.1 8 - 9 1422 71
XM_006229015.2 XP_006229077.1 8 - 9 1395 71
XM_006229016.2 XP_006229078.1 8 - 9 1283 71
XM_006229017.2 XP_006229079.1 8 - 9 1394 71
XM_006229018.2 XP_006229080.1 8 - 9 1421 71
XM_006229019.2 XP_006229081.1 8 - 9 1421 71
XM_006229020.2 XP_006229082.1 8 - 9 1394 71
GenBank mRNA AY346104.1 - 8 - 9 1360 71

View Full Product Details