Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
Slc35d3
Entrez Gene ID
308717
Gene Name
solute carrier family 35, member D3
Gene Aliases
-
Chromosome Location
Chr.1: 15298526 - 15301998 on Build Rnor_6.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001107522.1 NP_001100992.1 1 - 2 885 84

View Full Product Details