Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
Pak7
Entrez Gene ID
311450
Gene Name
p21 protein (Cdc42/Rac)-activated kinase 7
Gene Aliases
PAK-5, PAK-7
Chromosome Location
Chr.3: 129042241 - 129357491 on Build Rnor_6.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001107781.1 NP_001101251.1 3 - 4 558 153

View Full Product Details