Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
Gstcd
Entrez Gene ID
310855
Gene Name
glutathione S-transferase, C-terminal domain containing
Gene Aliases
RGD1311822
Chromosome Location
Chr.2: 238109758 - 238253287 on Build Rnor_6.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001107725.1 NP_001101195.1 3 - 4 1056 64
XM_006233324.2 XP_006233386.1 3 - 4 1137 64
XM_008761502.1 XP_008759724.1 2 - 3 1890 64
XM_008761503.1 XP_008759725.1 3 - 4 2713 64
XM_008761504.1 XP_008759726.1 3 - 4 2708 64
XM_008761505.1 XP_008759727.1 3 - 4 2671 64
GenBank mRNA FQ211741.1 - 3 - 4 1142 64

View Full Product Details