Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
Dkk2
Entrez Gene ID
295445
Gene Name
dickkopf WNT signaling pathway inhibitor 2
Gene Aliases
Dkk4
Chromosome Location
Chr.2: 237148918 - 237240883 on Build Rnor_6.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001106472.1 NP_001099942.1 2 - 3 1118 104

View Full Product Details