The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001077356.2 | NP_001070824.1 | 11 - 12 | 1990 | 148 |
GenBank mRNA | AB256513.1 | - | 11 - 12 | 1991 | 148 |